Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. p-3251536
Click to view available options
Quantity:
120 mL
500 mL
6 x 20 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 10μS/cm (10μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 5.0E-4
CAS Max % 100.0, 5.0E-4
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 120 mL
Concentration or Composition (by Analyte or Components) 4.7 ppm as NaCl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.