Learn More
Conductivity Standard, 45 mS/cm, Reagents
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard, 45 mS/cm |
| Packaging | Poly Bottle |
| CAS | 7732-18-5, 7647-14-5 |
| Specific Gravity | 1.0211 |
| Physical Form | Liquid |
| Molecular Formula | NaCl/H2O |
| Technique | Calibration of Conductivity Instrument |
| Color | Colorless |
| Boiling Point | 100°C |
| For Use With (Equipment) | Conductivity Meter |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.