Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
SDP

Catalog No. p-7239406 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2236024 120 mL
223602-16 500 mL
223602-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 2236024 Supplier Ricca Chemical Company Supplier No. 2236.024
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 5μS/cm (5μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 2.0E-4
CAS Max % 100.0, 2.0E-4
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 120 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.