Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Conductivity Standard, 100 μS/cm at 25°C (47.2 ppm TDS as NaCl), Ricca Chemical

Click to view available options
Quantity:
100 mL
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack
Specifications
Specifications
Chemical Name or Material | Conductivity/TDS Standard |
Packaging | Pack |
Name Note | 100μS/cm (100μmho/cm) at 25°C |
CAS | 7647-14-5, 7732-18-5 |
CAS Min % | 98.0, 0.0046 |
CAS Max % | 100.0, 0.0048 |
Physical Form | Liquid |
Color | Colorless |
pH | 6.7 to 7.3 |
Melting Point | 0°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction