Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 100 μS/cm at 25°C (47.2 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 22371 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 100μS/cm (100μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.0046
CAS Max % 100.0, 0.0048
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 4 L
Concentration or Composition (by Analyte or Components) 47.2ppm as NaCl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.