Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical
SDP

Catalog No. p-3251547 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

65,750 ppm TDS as NaCl

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 100mS/cm (100, 000μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 93.0, 4.85
CAS Max % 97.0, 5.05
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 65, 750ppm as NaCl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.