Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Conduritol B epoxide
GREENER_CHOICESDP

Catalog No. 70565 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-565 5 mg
70-562-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-565 Supplier Tocris Bioscience™ Supplier No. 7056/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Irreversible glucocerebrosidase (β-glucosidase) inhibitor; used to generate Gaucher disease models

Conduritol B epoxide is an irreversible glucocerebrosidase (GBA) inhibitor (Ki = 53 μM; IC50 = 4.28 - 9.49 μM). Also inhibitis lysosomal α-glucosidase. Used to generate animal models of Gaucher disease. Accelerates recovery of peripheral nerve injury in mice. Preserves motoneurons and ganglioside distribution at the neuromuscular junction, delays disease onset and improves motor function in a mouse ALS model.

Specifications

Chemical Name or Material DL-1,2-Anhydro-myo-Inositol
CAS 6090-95-5
Quantity 5 mg
Target Glycosylase Inhibitors
Molecular Formula C6H10O5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.