Learn More
Eisco™ Conglomerate Specimen
SCI_ED
Catalog No. S24723 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24723 12/Pk. 2 to 3 cm
S26617 10/Pk. 2 to 3 cm
S26625 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24723 Supplier Eisco™ Supplier No. ESNG0031PK12
Add to Cart
Add to Cart
Conglomerate Specimen has large clasts that indicate that pieces of rock underwent large amount of rolling, bouncing, and other weathering to become round.
  • Large clasts within rock are ideal for teaching students about rounding and sorting
  • For use in teaching students about sorting and rounding
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Well-rounded and poorly sorted, indicating it was deposited and formed in running water
Specifications
Product Type Sedimentary Rock
For Use With (Application) Lessons on depositional environments and identification of sedimentary rocks
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Conglomerate