Learn More
Tocris Bioscience™ Congo Red
Description
Specifications
Specifications
| Purity | >98% |
| For Use With (Application) | Fluorescent Amyloid B Probes |
| Quantity | 50 mg |
| Product Type | Congo Red |
| Molecular Weight (g/mol) | 696.66 |
| Inhibitors | Congo Red |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.