Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red Indicator, 0.1% (w/v), Ricca Chemical
SDP

Catalog No. p-3251568 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle

Specifications

Chemical Name or Material Congo Red Indicator
CAS 573-58-0, 7732-18-5
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Concentration 0.1% (w/v)
Percent Purity 0.1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Red
Quantity 120 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.