Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Congo Red TS, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Congo Red |
| Name Note | Test Solution |
| CAS | 7732-18-5, 64-17-5, 573-58-0, 67-63-0, 67-56-1 |
| CAS Min % | 90.0, 6.54, 0.45, 0.35 |
| CAS Max % | 94.0, 6.8, 0.47, 0.36 |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Color | Orange/Red |
| pH | 8 to 9.5 |
| Quantity | 120 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction