Learn More
United Scientific™ Conical Measuring Cylinders, Polypropylene

Shop All United Scientific Supplies, Inc Products


Description
- Our polypropylene conical measuring cylinders are ideal for measuring small quantities of reagents.
- They are autoclavable and have excellent contact clarity.
- The hexagonal base provides excellent stability to the cylinder, and the tapered spout enables easy pouring of reagents.
- Features easy-to-read raised graduations.
Specifications
Specifications
| Capacity (Metric) | 200 mL |
| Material | Polypropylene |
| Graduations | 5 mL |
| Base Type | Hexagonal |
| Height (English) | 5.8 in. |
| Height (Metric) | 149 mm |
| Product Type | Graduated Measuring Cylinder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.