Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Conical Measuring Cylinders, Polypropylene
SDPSCI_ED

Catalog No. S16055 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
12 mL
50 mL
125 mL
200 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S16055 12 mL
S16003 200 mL
S16075 50 mL
S15792 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S16055 Supplier United Scientific™ Supplier No. 81150
Add to Cart
Add to Cart

Unique design provides users with a different option than standard cylinders.

  • Our polypropylene conical measuring cylinders are ideal for measuring small quantities of reagents.
  • They are autoclavable and have excellent contact clarity.
  • The hexagonal base provides excellent stability to the cylinder, and the tapered spout enables easy pouring of reagents.
  • Features easy-to-read raised graduations.

Specifications

Capacity (Metric) 12 mL
Material Polypropylene
Graduations 1 mL
Base Type Hexagonal
Height (English) 2.55 in.
Height (Metric) 65 mm
Product Type Graduated Measuring Cylinder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.