Learn More
United Scientific™ Connecting Cord Organizer
SDPSCI_ED
Catalog No. S03039 Shop All United Scientific Supplies, Inc Products
missing translation for 'changeView'
Click to view available options
Includes:
No Stand
Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S03039 No Stand
S03038 Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Prop 65
Catalog No. S03039 Supplier United Scientific™ missing translation for 'supplierNo' WRHLDR1
Only null left
Add to cart
Add to cart
Prop 65
Safely and neatly organize connecting cords in your classroom with Connecting Cord Organizer.
Many electricity lessons and labs require alligator or banana plug connecting cords. Often these cords are stored in a box, and can be difficult to untangle. The Connecting Cord Organizer easily holds multiple wires. The organizer mounts to a standard support stand, and includes 14 holes that fit banana plug cords and 6 hooks that can be used to loop or clip alligator clip wires. The Connecting Cord Organizer requires a support stand with a rod diameter of 3/8 inch or smaller. The Connecting Cord Organizer with Stand includes a 5 x 8 inch cast iron support stand with a 20 inch rod.
  • Holds 14 Banana plug cords and approximately 18 alligator clip cords
  • Works with support rods up to 3/8 in. in diameter
  • Holds wires securely and within easy reach
  • Powder black coating
Specifications
Product Type Connecting Wires
For Use With (Equipment) Power Supplies
Includes No Stand