Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Connectors for Milli-Q™ IQ 7000 Ultrapure Water System
GSA_VA

Catalog No. p-7120929
Click to view available options
Type:
Connector 2m Q-POD to Q-POD
Connector 2m System to Q-POD
Connector 5m Q-POD to Q-POD
Connector 5m System to Q-POD

The most integrated pure and ultrapure water system that is intelligent, intuitive and green.

Order Info

Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Type Connector 2m Q-POD to Q-POD
Purity Meter Included No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.