Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 25/GJB7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Connexin 25/GJB7 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Connexin 25/GJB7 Polyclonal antibody specifically detects Connexin 25/GJB7 in Human samples. It is validated for ImmunofluorescenceSpecifications
Connexin 25/GJB7 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
bA136M9.1, connexin 25, connexin25, Connexin-25, Cx25, CX25connexin-25, gap junction beta-7 protein, gap junction protein, beta 7, gap junction protein, beta 7, 25kDa, MGC71746 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: YKLYDGFSVPYLIKCDLKPCPNTVDCFISKPTEKTIFI | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
375519 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title