Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 26/GJB2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB201682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201682 Supplier Novus Biologicals Supplier No. NBP241304AF488
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Connexin 26/GJB2 Polyclonal antibody specifically detects Connexin 26/GJB2 in Human, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Connexin 26/GJB2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias beta 2, 26kD (connexin 26), Cx26, CX26DFNA3, DFNA3A, DFNB1, gap junction protein, beta 2, 26kDa, gap junction protein, beta 2, 26kDa (connexin 26), NSRD1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human GJB2. The immunogen is located within amino acids 80 - 130 of GJB2. Amino Acid Squence: HVAYRRHEKKRKFIKGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Sensory Systems
Primary or Secondary Primary
Gene ID (Entrez) 2706
Target Species Human, Rat, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.