Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin-29 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988712
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8712 Supplier Bioss Supplier No. BS8697R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen Connexin-29
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GJC3
Gene Accession No. Q8NFK1
Gene Alias connexin 29; Connexin-29; Connexin-30.2; Connexin-31.3; Cx29; Cx30.2; Cx31.3; Gap junction epsilon-1 protein; Gap junction gamma-3 protein; gap junction membrane channel protein epsilon 1; gap junction protein gamma 3; gap junction protein, gamma 3; gap junction protein, gamma 3, 30.2kDa; Gjc3; Gje1
Gene Symbols GJC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Connexin 29.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349149
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.