Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 30.3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565923 Supplier Invitrogen™ Supplier No. PA565923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLYKDYDMPRV VACSVEPCPH TVDCYISR Highest antigen sequence identity to the following orthologs - mouse 86%, rat 90%.

This gene encodes a transmembrane connexin protein that is a component of gap junctions. Mutations in this gene have been associated with erythrokeratodermia variabilis, progressive symmetric erythrokeratoderma and hearing impairment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 30.3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GJB4
Gene Accession No. Q9NTQ9
Gene Alias Cnx30.3; connexin 30.3; connexin-30.3; CX30.3; Cxn-30.3; EKV; gap junction beta-4 protein; gap junction membrane channel protein beta 4; gap junction protein beta 4; gap junction protein, beta 4; gap junction protein, beta 4, 30.3kDa; Gjb4; Gjb-4
Gene Symbols GJB4
Host Species Rabbit
Immunogen Recombinant Human Connexin 30.3. Recombinant protein control fragment (Product #RP-105008).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127534
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.