Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 30 Monoclonal Antibody (CL4540)
GREENER_CHOICE

Catalog No. PIMA535021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535021 Supplier Invitrogen™ Supplier No. MA535021
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: RSKRAQTQKN HPNHALKESK Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 85%.

Gap junctions allow the transport of ions and metabolites between the cytoplasm of adjacent cells. They are formed by two hemichannels, made up of six connexin proteins assembled in groups. Each connexin protein has four transmembrane segments, two extracellular loops, a cytoplasmic loop formed between the two inner transmembrane segments, and the N- and C-terminus both being in the cytoplasm. The specificity of the gap junction is determined by which connexin proteins comprise the hemichannel. In the past, connexin protein names were based on their molecular weight, however the new nomenclature uses sequential numbers based on which form of the gap junction is present. This gene encodes one of the connexin proteins. Mutations in this gene have been found in some forms of deafness and in some families with hidrotic ectodermal dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 30
Applications Immunocytochemistry
Classification Monoclonal
Clone CL4540
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GJB6
Gene Accession No. O95452
Gene Alias AA958971; connexin 30; connexin-30; CX30; Cxn-30; D14Bwg0506e; DFNA3; DFNA3B; DFNB1B; ECTD2; ectodermal dysplasia 2, hidrotic (Clouston syndrome); ED2; EDH; Gap junction beta-6 protein; gap junction membrane channel protein beta 6; gap junction protein beta 6; gap junction protein, beta 6; gap junction protein, beta 6 (connexin 30); gap junction protein, beta 6, 30kDa; Gjb6; HED; HED2
Gene Symbols GJB6
Host Species Mouse
Immunogen Synthetic Peptide corresponding to Human Connexin 30.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.