Learn More
Invitrogen™ Connexin 31.1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total GJB5.
Specifications
Specifications
| Antigen | Connexin 31.1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Gjb5 |
| Gene Accession No. | O95377, P28232, Q02739 |
| Gene Alias | Cnx31.1; connexin 31.1; connexin-31.1; Cx31.1; Cxn-31.1; Gap junction beta-5 protein; gap junction membrane channel protein beta 5; gap junction protein beta 5; gap junction protein, beta 5; gap junction protein, beta 5 (connexin 31.1); gap junction protein, beta 5, 31.1kDa; gap junctional protein cx31.1; Gjb5; Gjb-5 |
| Gene Symbols | Gjb5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.