Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 32 Monoclonal Antibody (CX-2C2), Abbomax

Catalog No. 502204375
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4375 Supplier Abbomax Inc Supplier No. 605650
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: A549 or Heart Cellular location: Cell membrane, Cell junction.

Connexin 32 Monoclonal antibody specifically detects Connexin 32 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Connexin 32
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CX-2C2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene GJB1
Gene Accession No. P08034
Gene Alias CMTX; CMTX1; Cnx32; connexin 32; connexin-32; CX32; Cxn-32; GAP junction 28 kDa liver protein; gap junction beta-1 protein; gap junction membrane channel protein beta 1; gap junction protein beta 1; gap junction protein, beta 1; gap junction protein, beta 1, 32kDa; Gjb1
Host Species Mouse
Immunogen A synthetic peptide of human Connexin 32
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2705
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.