Learn More
Invitrogen™ Connexin 36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 516300

Description
Connexin 36 Polyclonal Antibody for Western Blot, IP, ELISA
Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.
Specifications
Connexin 36 | |
Polyclonal | |
Unconjugated | |
GJD2 | |
connexin 36; connexin36; connexin-36; cx36; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; Gja9; GJD2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14617, 50564, 57369 | |
-20°C | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
O54851, O70610, Q9UKL4 | |
GJD2 | |
Peptide corresponding to a sequence from the C-terminus of rat and mouse Connexin 36 (Cx36). This sequence differs from the human Cx36 protein by a single amino acid. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.