Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 364600
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-460-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-460-0 Supplier Invitrogen™ Supplier No. 364600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with mouse Connexin 36. On western blots, it identifies a single band at ~36 kDa. The specificity of this antibody was demonstrated using Connexin 36 knockout mice. Reactivity has been confirmed with mouse Connexin 36 transfected HeLa cell lysates, mouse brain, retina and pancreas homogenates.

Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 36
Applications ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJD2
Gene Accession No. O54851, Q9UKL4
Gene Alias connexin 36; connexin36; connexin-36; cx36; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; Gja9; GJD2
Gene Symbols GJD2
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human Connexin 36 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14617, 57369
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.