Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin-36 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2376 Supplier Bioss Supplier No. BS1266R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.

Specifications

Antigen Connexin-36
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GJD2
Gene Accession No. Q9UKL4
Gene Alias connexin 36; connexin36; Connexin-36; Cx36; Gap junction alpha-9 protein; Gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; GJA9; GJD2
Gene Symbols GJD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Connexin-36 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57369
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.