Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 43/GJA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300309
Description
Connexin 43/GJA1 Polyclonal specifically detects Connexin 43/GJA1 in Rat, Avian samples. It is validated for Western Blot, Immunohistochemistry (Negative).Specifications
Connexin 43/GJA1 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry (Negative) | |
P08050 | |
GJA1 | |
Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # P08050 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼43k connexin43 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
Rabbit | |
43 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
2697 | |
Rat, Avian | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction