Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 43/GJA1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151938
Description
Connexin 43/GJA1 Polyclonal specifically detects Connexin 43/GJA1 in Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
Connexin 43/GJA1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human and Rat. Expected from sequence similarity: Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1:100-1:2000, Peptide ELISA Detection limit 1:64000 | |
NP_000156.1 | |
GJA1 | |
Peptide with sequence C-QPFDFPDDNQNSKK corresponding to internal region according to NP_000156.1. | |
0.1 mg | |
Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
2697 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction