Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 43 Mouse anti-Human, Mouse, Rat, Clone: P4G9, MilliporeSigma™

Catalog No. MABT901MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT901MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT901MI Supplier MilliporeSigma Supplier No. MABT901
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Connexin 43, C-terminal, clone P4G9 Immunofluorescence, Immunohistochemistry (Paraffin), and Immunoprecipitation for the detection of Connexin-43.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 43
Applications Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone P4G9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Gja1,Cx43
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal region of rat connexin-43.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_036699
Test Specificity Clone P4G9 specifically detects connexin 43 in human and rat heart. It targets an epitopw within 23 amino acids from the C-terminal end.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.