Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin-46 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733932 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3932 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3932 Supplier Proteintech Group Inc Supplier No. 227171AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Connexin 46 (Cx46), also known as Gap Junction Protein Alpha-3 (GJA3), CAE3, CX46 and C2P3, maps to human chromosome 13q11-q12 and encodes a 46 kDa protein. Cx46, along with Cx50, is principally expressed in the lens of the eye. Cx46 mediates intercellular interactions during development and is necessary for the survival of neural crest cells. Cx46 forms gap junctions that connect lens fiber cells and are crucial for maintaining lens transparency and normal lens function. Mutations of Cx46 result in severe cataracts of the lens. Individual knockouts of Cx46 and Cx50 lead to changes in the rate of lens fiber cell differentiation and cell size, thus the interaction of Cx46 and Cx50 is required for proper organization of fiber cells.

Specifications

Antigen Connexin-46
Applications Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GJA3
Gene Accession No. Q9Y6H8
Gene Alias Connexin-46, CX46, CZP3, GJA3
Gene Symbols GJA3
Host Species Rabbit
Immunogen Connexin-46 Fusion Protein Ag18580
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2700
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.