Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 58/GJA9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | Connexin 58/GJA9 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Connexin 58/GJA9 Polyclonal specifically detects Connexin 58/GJA9 in Human samples. It is validated for Western Blot.Specifications
| Connexin 58/GJA9 | |
| Polyclonal | |
| Rabbit | |
| P57773 | |
| 81025 | |
| Synthetic peptides corresponding to GJA9(gap junction protein, alpha 9, 59kDa) The peptide sequence was selected from the middle region of GJA9. Peptide sequence IDGENNMRQSPQTVFSLPANCDWKPRWLRATWGSSTEHENRGSPPKGNLK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| connexin 59, connexin-58, Connexin-59, CX58, Cx59, CX59gap junction alpha 10, Gap junction alpha-10 protein, gap junction alpha-9 protein, gap junction protein, alpha 10, 59kDa, gap junction protein, alpha 9, 59kDa, GJA10, MGC50985 | |
| GJA9 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title