Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Consolidator™ Nitrile Coated Work Gloves with Interlock Cotton Liner

Catalog No. p-7230396 Shop All MCR Safety Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a full nitrile coating for abrasion resistance with a seamless palm for increased comfort and dexterity.

  • Cut and sewn into a snug pattern to allow for maximum dexterity
  • Full nitrile coating for abrasion resistance
  • Slip-on cuff for easy donning and doffing
  • Straight thumb
  • Clute cut

Specifications

Construction Cut and Sewn
Coating Material Nitrile
Color Blue
Exterior Finish No Texture
Grade Industrial
Hand Specific
Material Nitrile
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Automotive Assembly and Carpentry
Glove Cuff Style Open
Liner Material Cotton
Series 9700
Thumb Style Straight
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.