Learn More
Werner Co. Construction/Maintenance Kit
Description
Contains common components for a safe fall protection system for a variety of environments and a range of applications.
- Includes BaseWear harness with pass-through leg straps, 6 ft. SoftCoil lanyard, and a cross arm strap for a basic anchor.
- Packaged in waterproof bucket.
Specifications
Specifications
| Size | Medium/Large |
| Material | Polyester |
| Buckle Style (Leg) | Slotted Pass-through |
| Webbing Material | Polyester |
| For Use With (Application) | Fall Protection |
| Product Type | Climbing Safety Harness |
| Color | Black, Blue, Yellow |
| D-Ring Location | Back |
| Includes | BaseWear Harness H411002 with Pass-through Leg Straps, 6 ft. SoftCoil Lanyard C351100, and a Cross Arm Strap A111106 for a Basic Anchor |
| Style | Aerial/Construction/ Maintenance Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.