Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Contact Dishes
Shop All Greiner Bio-One Products
Click to view available options
Vent:
No
Yes
Description
- Available in different sizes
- With or without vents
- Material: Crystal clear and heat resistant polystyrene
- Contact dish with graduation to enable quick and easy analyses
Specifications
Specifications
| Color | Clear |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Cell Culture Dish |
| Vent | No |
| Height (Metric) | 15 mm |
| Diameter (Metric) | 65 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction