Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Contact Plates
SCI_ED

Catalog No. S012396
Click to view available options
Media Type:
Levine EMB Agar
Potato Dextrose Agar
SDA with Lecithin and Tween 80
TSA w/ Lecithin and Tween 80
Tryptic Soy Agar
Detects:
Bacteria
Bacteria and Fungi
Fungi
Gram Negative Bacteria
Microorganisms
Prop 65: Cancer and Reproductive Harm.
Prop 65: Cancer and Reproductive Harm.

Contact plates ideal for testing surfaces.

  • Available in a variety of formulations
  • For environmental monitoring of surfaces
  • Steam sterilized
  • Agar is slightly raised to make it easy to test surfaces

Specifications

Detects Bacteria and Fungi
Format Contact Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type Potato Dextrose Agar
Product Type Contact Plate
Sterility Sterile
Target Organism Class Fungi, Bacteria, Yeast, Molds
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 10 Plates
Dimensions (Dia. x H) 65 x 15 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.