Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Contact Plates
Shop All Fischer Technical Company Products

Click to view available options
Media Type:
Levine EMB Agar
Potato Dextrose Agar
SDA with Lecithin and Tween 80
TSA w/ Lecithin and Tween 80
Tryptic Soy Agar
Detects:
Bacteria
Bacteria and Fungi
Fungi
Gram Negative Bacteria
Microorganisms
Description
- Available in a variety of formulations
- For environmental monitoring of surfaces
- Steam sterilized
- Agar is slightly raised to make it easy to test surfaces
Specifications
Specifications
| Detects | Gram Negative Bacteria |
| Format | Contact Plate |
| Manufacturing Origin | US with Globally Sourced Materials |
| Media Type | Levine EMB Agar |
| Product Type | Contact Plate |
| Sterility | Sterile |
| Target Organism Class | Gram-negative Bacteria |
| Test Type | Microbiological Tests |
| For Use With (Application) | Microbiological Testing |
| Quantity | 10 Plates |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction