Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299275R
Description
Contactin-1 Polyclonal specifically detects Contactin-1 in Human samples. It is validated for ELISA.Specifications
Contactin-1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
1272 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 550 | |
contactin 1, contactin-1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
Produced in rabbits immunized with purified, recombinant Contactin-1 (NP_001834.2; Met 1-993). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction