Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 1 Monoclonal Antibody (10B3B1D1)
GREENER_CHOICE

Catalog No. PIMA529177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529177 Supplier Invitrogen™ Supplier No. MA529177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CNTN1/Contactin-1.

Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 1
Applications ELISA
Classification Monoclonal
Clone 10B3B1D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Cntn1
Gene Accession No. Q12860
Gene Alias AW495098; CNTN; CNTN1; contactin 1; contactin-1; F3; F3cam; glycoprotein gP135; GP135; MYPCN; neural cell surface protein F3
Gene Symbols Cntn1
Host Species Mouse
Immunogen Recombinant Human CNTN1 protein (Met1-Ser993).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.