Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Contactin 1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547959
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Contactin 1 | |
Polyclonal | |
Unconjugated | |
Cntn1 | |
AW495098; CNTN; CNTN1; contactin 1; contactin-1; F3; F3cam; glycoprotein gP135; GP135; MYPCN; neural cell surface protein F3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
117258, 1272, 12805 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P12960, Q12860, Q63198 | |
Cntn1 | |
Sf 21-derived recombinant human Contactin-1 Glu21-Ser993. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction