Learn More
Invitrogen™ Contactin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121133
Description
Positive test controls include: Mouse brain, Mouse spinal cord, Rat brain. The target is usually found in the following locations: Cell membrane, GPI-anchor, Lipid-anchor. Immunogen sequence: EVKIRSYNRR GDGPESLTAL VYSAEEEPRV APTKVWAKGV SSSEMNVTWE PVQQDMNGIL LGYEIRYWKA GDKEAAADRV RTAGLDTSAR VSGLHPNTKY HVTVRAYNRA GTGPASPSAN ATTMKPPPRR PPGNISWTFS SSSLSIKWDP VVPFRNESAV TGYKMLYQND LHLTPTLHLT GKNWIEIPVP EDIGHALVQI RTTGPGGDGI PAEVHIVRNG GTSMMVEN.
Contactin 2 (CNTN2, transiently-expressed axonal glycoprotein, AXT, TAX, TAX1, TAG-1, axonin 1) is a neuronal cell adhesion molecule (CAM) that influences the formation of axon connections in the developing nervous system. Contactin 2 is a member of the immunoglobulin superfamily (IgSF) and contains a glycosylphosphatidylinositol-anchor, six immunoglobulin (Ig)-like and four Fibronectin type III (FNIII)-like domains. Contactin 2 is expressed predominantly during neural development on the cell membrane of axons in nerve fiber tracts in order to guide commissural axons without promoting their growth. Contactin 2 binds with NgCAM in the plane of the same membrane (cis-binding). The Contactin 2 heterophilic (Contactin 2/NgCAM and Contactin 2/NrCAM) binding sites are localized to the first four Ig domains. The Contactin 2 homophilic (Contactin 2/Contactin 2) binding site is localized to the FNIII domain.
Specifications
Contactin 2 | |
Polyclonal | |
Unconjugated | |
CNTN2 | |
Axonal glycoprotein TAG-1; axonin; Axonin 1; Axonin1; axonin-1; axonin-1 cell adhesion molecule; AXT; Cntn2; contactin 2; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin2; contactin-2; D130012K04Rik; DKFZp781D102; FAME5; MGC157722; TAG1; TAG-1; TAG-564; Tax; TAX1; TAX-1; transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21367, 25356 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22063, Q61330 | |
CNTN2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 785-1012 of human Contactin 2 (NP_005067.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.