Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 2 Recombinant Rabbit Monoclonal Antibody (4R2S3)
GREENER_CHOICE

Catalog No. PIMA542873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542873 Supplier Invitrogen™ Supplier No. MA542873
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: A-549, C6, SH-SY5Y, U-251MG, Mouse lung, Mouse spinal cord, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, GPI-anchor, Lipid-anchor. Immunogen sequence: MGTATRRKPH LLLVAAVALV SSSAWSSALG SQTTFGPVFE DQPLSVLFPE ESTEEQVLLA CRARASPPAT YRWKMNGTEM KLEPGSRHQL VGGNLVIMNP.

Contactin 2 (CNTN2, transiently-expressed axonal glycoprotein, AXT, TAX, TAX1, TAG-1, axonin 1) is a neuronal cell adhesion molecule (CAM) that influences the formation of axon connections in the developing nervous system. Contactin 2 is a member of the immunoglobulin superfamily (IgSF) and contains a glycosylphosphatidylinositol-anchor, six immunoglobulin (Ig)-like and four Fibronectin type III (FNIII)-like domains. Contactin 2 is expressed predominantly during neural development on the cell membrane of axons in nerve fiber tracts in order to guide commissural axons without promoting their growth. Contactin 2 binds with NgCAM in the plane of the same membrane (cis-binding). The Contactin 2 heterophilic (Contactin 2/NgCAM and Contactin 2/NrCAM) binding sites are localized to the first four Ig domains. The Contactin 2 homophilic (Contactin 2/Contactin 2) binding site is localized to the FNIII domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4R2S3
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNTN2
Gene Accession No. Q02246, Q61330
Gene Alias Axonal glycoprotein TAG-1; axonin; Axonin 1; Axonin1; axonin-1; axonin-1 cell adhesion molecule; AXT; Cntn2; contactin 2; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin2; contactin-2; D130012K04Rik; DKFZp781D102; FAME5; MGC157722; TAG1; TAG-1; TAG-564; Tax; TAX1; TAX-1; transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein
Gene Symbols CNTN2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21367, 6900
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.