Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-2/TAG1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298381JF646
Description
Contactin-2/TAG1 Polyclonal antibody specifically detects Contactin-2/TAG1 in Mouse samples. It is validated for ELISASpecifications
Contactin-2/TAG1 | |
Polyclonal | |
ELISA | |
Axonal glycoprotein TAG-1, Axonin-1, axonin-1 cell adhesion molecule, AXT, contactin 2 (axonal), contactin 2 (transiently expressed), contactin-2, DKFZp781D102, FLJ37193, MGC157722, TAG1, TAG-1, TAX, TAX-1, TAX1FLJ42746, Transient axonal glycoprotein 1, transiently-expressed axonal glycoprotein | |
Produced in rabbits immunized with purified, recombinant Mouse Contactin-2/TAG1 (Uniprot#: Q61330; Gln31-Glu1013) | |
0.1 mL | |
Cancer, Cell Biology, Immunology, Neuroscience | |
6900 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction