Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-3 Antibody (524518), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6644MFV450
Description
Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Contactin-3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5067 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
524518 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
contactin 3 (plasmacytoma associated) | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3, Glu20-Thr1003, Accession # Q07409 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction