Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 3 Monoclonal Antibody (3B9A1E2), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA542980
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542980 100 μL
PIMA543515 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA542980 Supplier Invitrogen™ Supplier No. MA542980
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human CNTN3/Contactin-3/BIG-1

Contactin 3 is a type of contactin that mediate cell surface interactions during nervous system development. Has some neurite outgrowth-promoting activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 3
Applications ELISA
Classification Monoclonal
Clone 3B9A1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CNTN3
Gene Accession No. Q9P232
Gene Alias BIG-1; Brain-derived immunoglobulin superfamily protein 1; Cntn3; contactin 3; contactin 3 (plasmacytoma associated); contactin-3; KIAA1496; Pang; PCS; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1)
Gene Symbols CNTN3
Host Species Mouse
Immunogen Recombinant Human CNTN3 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.