Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-4 Antibody (SR1190), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Contactin-4 |
---|---|
Clone | SR1190 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Contactin-4 Monoclonal antibody specifically detects Contactin-4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Contactin-4 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
A synthesized peptide derived from human Contactin-4 (Uniprot #: Q8IWV2) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1190 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
152330 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title