Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587824 Supplier Invitrogen™ Supplier No. PA587824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGVFNNKGEG PFSPTTVVYS AEEEPTKPPA SIFARSLSAT DIEVFWASPL EKNRGRIQGY EVKYWRHEDK EENARKIRTV GNQTSTKITN LKGSVLYHLA VKAYNSAGTG PSSATVNVTT RKPPPSQPPG NIIWNSSDSK IILNWDQVKA LDNESEVKGY KVLYRWNRQS STSVIETNKT SVELSLPFDE DYIIEIKPFS DGGDGSSSEQ IRIPKISNAY ARGSGASTSN ACTLSAISTI MISLTARSSL; Positive Samples: U-87MG, HeLa, HT-1080; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.

The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNTN4
Gene Accession No. Q8IWV2
Gene Alias 9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16
Gene Symbols CNTN4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 448-697 of human CNTN4 (NP_0011938851).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152330
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.