Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-5 Antibody (024), Alexa Fluor™ 750 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289278AF750
Description
Contactin-5 Monoclonal antibody specifically detects Contactin-5 in Human samples. It is validated for ELISA, ELISASpecifications
Contactin-5 | |
Monoclonal | |
Alexa Fluor 750 | |
contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-5 (NP_055176.1; Met1-Gln1059). | |
0.1 mL | |
Neuroscience | |
53942 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
024 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction