Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin-5 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299347F
Description
Contactin-5 Polyclonal specifically detects Contactin-5 in Human samples. It is validated for ELISA.Specifications
Contactin-5 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
53942 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
Produced in rabbits immunized with purified, recombinant Contactin-5 (NP_055176.1; Met 1-Gln 1059). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction