Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Contactin 6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589720
Description
Immunogen sequence: DGLLSRPIFT QEPHDVIFPL DLSKSEVILN CAANGYPSPH YRWKQNGTDI DFTMSYHYRL DGGSLAINSP HTDQDIGMYQ CLATNLLGTI L; Positive Samples: U-87MG, Jurkat; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.
The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
Specifications
Contactin 6 | |
Polyclonal | |
Unconjugated | |
CNTN6 | |
CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27255 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.53 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9UQ52 | |
CNTN6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-110 of human CNTN6 (NP_0552761). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction