Learn More
Description
- Ready to use swab
- Assay of microbial load directly from surfaces
- Kit contents: 30 sealed envelopes (swab + culture medium) and one instruction sheet
- Watch for the color change of the medium and interpret the results
Specifications
Specifications
| Detection Method | Rapid Detection |
| Sample Type | Environmental Sample |
| For Use With (Application) | Work Surface Contamination Monitoring |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
