Learn More
Description
- Ready to use swab with chromogenic culture medium
- Kit contents: 30 sealed envelopes (swab + culture medium) and one instruction sheet
- Watch for the color change of the medium, the emission of blue fluorescence under Wood's lamp and interpret the results
Specifications
Specifications
| Sample Type | Environmental Sample |
| Detection Method | Rapid Detection |
| For Use With (Application) | Work Surface Contamination Monitoring |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
